missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human USP28 Partial ORF (NP_065937, 978 a.a. - 1077 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_065937 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 57646 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16112873
|
Abnova™
H00057646-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16102873
|
Abnova™
H00057646-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
The ubiquitin-dependent protein degradation pathway is essential for proteolysis of intracellular proteins and peptides. Enzymes that remove ubiquitin from ubiquitin-conjugated peptides, like USP28, affect the fate and degradation of intracellular proteins and are essential for maintenance of cell-free ubiquitin pools (Valero et al., 2001).[supplied by OMIM]
Sequence: VMNELIIPCIHLIINNDISKDDLDAIEVMRNHWCSYLGQDIAENLQLCLGEFLPRLLDPSAEIIVLKEPPTIRPNSPYDLCSRFAAVMESIQGVSTVTVKSpecifications
NP_065937 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1515 | |
USP28 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
57646 | |
USP28 (Human) Recombinant Protein (Q01) | |
VMNELIIPCIHLIINNDISKDDLDAIEVMRNHWCSYLGQDIAENLQLCLGEFLPRLLDPSAEIIVLKEPPTIRPNSPYDLCSRFAAVMESIQGVSTVTVK | |
RUO | |
USP28 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |