Learn More
Abnova™ Human USP21 Partial ORF (NP_036607, 466 a.a. - 565 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00027005-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the C19 peptidase family, also known as family 2 of ubiquitin carboxy-terminal hydrolases. The encoded protein cleaves ubiquitin from ubiquitinated proteins for recycling in intracellular protein degradation. The encoded protein is also able to release NEDD8, a ubiquitin-like protein, from NEDD8-conjugated proteins. This gene has been referred to as USP16 and USP23 but is now known as USP21. Alternatively spliced transcript variants have been described. [provided by RefSeq]
Sequence: FSASRGSIKKSSVGVDFPLQRLSLGDFASDKAGSPVYQLYALCNHSGSVHYGHYTALCRCQTGWHVYNDSRVSPVSENQVASSEGYVLFYQLMQEPPRCLSpecifications
NP_036607 | |
Liquid | |
27005 | |
USP21 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC3394/USP16/USP23 | |
USP21 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FSASRGSIKKSSVGVDFPLQRLSLGDFASDKAGSPVYQLYALCNHSGSVHYGHYTALCRCQTGWHVYNDSRVSPVSENQVASSEGYVLFYQLMQEPPRCL | |
RUO | |
USP21 | |
Wheat Germ (in vitro) | |
GST |