missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human USP12 Partial ORF (NP_872294, 69 a.a. - 168 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00219333-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16102447

  • 335.00€ / 10µg

Estimated Shipment: 11-05-2023
to see stock.
Add to basket

Description

Description

Sequence: AYKSQPRKKESLLTCLADLFHSIATQKKKVGVIPPKKFITRLRKENELFDNYMQQDAHEFLNYLLNTIADILQEERKQEKQNGRLPNGNIDNENNNSTPD
Specifications

Specifications

NP_872294
Liquid
219333
USP12 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
USP12L1
USP12
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AYKSQPRKKESLLTCLADLFHSIATQKKKVGVIPPKKFITRLRKENELFDNYMQQDAHEFLNYLLNTIADILQEERKQEKQNGRLPNGNIDNENNNSTPD
RUO
USP12
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers