missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human UBE2O Partial ORF (NP_071349.2, 361 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00063893-Q01.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16015615

  • 335.00€ / 10µg
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

Sequence: VEPAKIAWECPEKNCAQGEGSMAKKVKRLLKKQVVRIMSCSPDTQCSRDHSMEDPDKKGESKTKSEAESASPEETPDGSASPVEMQDEGAEEPHEAGEQLPPFLLKEGRD
Specifications

Specifications

NP_071349.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VEPAKIAWECPEKNCAQGEGSMAKKVKRLLKKQVVRIMSCSPDTQCSRDHSMEDPDKKGESKTKSEAESASPEETPDGSASPVEMQDEGAEEPHEAGEQLPPFLLKEGRD
RUO
UBE2O
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
63893
UBE2O (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
E2-230K/FLJ12878/KIAA1734
UBE2O
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers