missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human UAP1 Partial ORF (NP_003106, 406 a.a. - 504 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006675-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16125755

  • 359.00€ / 10µg

Estimated Shipment: 27-09-2022
to see stock.
Add to basket

Description

Description

Sequence: LKNADSQNGKDNPTTARHALMSLHHCWVLNAGGHFIDENGSRLPAIPRLKDANDVPIQCEISPLISYAGEGLESYVADKEFHAPLIIDENGVHELVKNG
Specifications

Specifications

NP_003106
Liquid
6675
UAP1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AGX1/AgX/SPAG2
UAP1
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LKNADSQNGKDNPTTARHALMSLHHCWVLNAGGHFIDENGSRLPAIPRLKDANDVPIQCEISPLISYAGEGLESYVADKEFHAPLIIDENGVHELVKNG
RUO
UAP1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers