Learn More
ACROBiosystems Human TYRO3 / Dtk Protein, Fc Tag
Brand: ACROBiosystems TY3-H5251-200UG
Additional Details : Weight : 0.00001kg
Description
Tyrosine-protein kinase receptor TYRO3 is also known as Tyrosine-protein kinase BYK, DTK, RSE, SKY, TIF, which belongs to the protein kinase superfamily, Tyr protein kinase family and AXL/UFO subfamily. TYRO3 regulates many physiological processes including cell survival, migration and differentiation. TYRO3 activates the AKT survival pathway, including nuclear translocation of NF-kappa-B and up-regulation of transcription of NF-kappa-B-regulated genes. TYRO3 interacts (via N-terminus) with extracellular ligands TULP1 and GAS6 By similarity and also interacts with PIK3R1, this interaction increases PI3-kinase activity.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 68.2 kDa. The protein migrates as 86-106 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
AAH51756.1 (Q261H) | |
200 μg | |
Ala 41 - Ser 428 | |
TYRO3,BYK,DTK,RSE,SKY,TIF | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
68.2 kDa | |
Human TYRO3, Fc Tag (TY3-H5251) is expressed from human 293 cells (HEK293). It contains AA Ala 41 - Ser 428 (Accession # AAH51756.1 (Q261H)). | |
-20°C | |
TYRO3 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |