missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TXNDC Full-length ORF (NP_110382.2, 1 a.a. - 280 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00081542-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195113

  • 349.00€ / 10µg

Estimated Shipment: 10-11-2023
to see stock.
Add to basket

Description

Description

TXNDC1 is a thioredoxin (TXN; see MIM 187700)-related protein with disulfide reductase activity (Matsuo et al., 2001 [PubMed 11152479]).[supplied by OMIM]

Sequence: MAPSGSLAVPLAVMVPLLWGAPWTHGRRSNVRVITDENWRELLEGDWMIEFYAPWCPACQNLQPEWESFAEWGEDLEVNIAKVDVTEQPGLSGRFIINALPTIYHCKDGEFRRYQGPRTKKDFINFISDKEWKSIEPVSSWFGPGSVLMSSMSALFQLSMWIRTCHNYFIEDLGLPVWGSYTVFALATLFSGLLLGLCMIFVADCLCPSKRRRPQPYPYPSKKLLSESAQPLKKVEEEQEADEEDVSEEEAESKEGTNKDFPQNAIRQRSLGPSLATDKS
Specifications

Specifications

NP_110382.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
58.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp564E1962/TMX/TXNDC
TXNDC1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
81542
TXNDC (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAPSGSLAVPLAVMVPLLWGAPWTHGRRSNVRVITDENWRELLEGDWMIEFYAPWCPACQNLQPEWESFAEWGEDLEVNIAKVDVTEQPGLSGRFIINALPTIYHCKDGEFRRYQGPRTKKDFINFISDKEWKSIEPVSSWFGPGSVLMSSMSALFQLSMWIRTCHNYFIEDLGLPVWGSYTVFALATLFSGLLLGLCMIFVADCLCPSKRRRPQPYPYPSKKLLSESAQPLKKVEEEQEADEEDVSEEEAESKEGTNKDFPQNAIRQRSLGPSLATDKS
RUO
TXNDC1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers