missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TSPAN9 Full-length ORF (AAH34280.1, 1 a.a. - 157 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH34280.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10867 |
Molecular Weight (g/mol) | 43.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16135772
|
Abnova™
H00010867-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16125772
|
Abnova™
H00010867-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. [provided by RefSeq]
Sequence: MWGRPTFEQSCPALVPWPGFLVRSFSRLRRNPQSADSGADTSGRRDSADKCCSCTVGPGASCVFCCGWGGWVGLCLSMQFLIFVNINSKSLVHWEMCNLPENLFCFWSTSGVASGPRAFATVLPPAPTSSVCLQSLIYRSPRCLLYSLCAWPFCYLASpecifications
AAH34280.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
43.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MWGRPTFEQSCPALVPWPGFLVRSFSRLRRNPQSADSGADTSGRRDSADKCCSCTVGPGASCVFCCGWGGWVGLCLSMQFLIFVNINSKSLVHWEMCNLPENLFCFWSTSGVASGPRAFATVLPPAPTSSVCLQSLIYRSPRCLLYSLCAWPFCYLA | |
RUO | |
TSPAN9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10867 | |
TSPAN9 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NET-5/PP1057 | |
TSPAN9 | |
Recombinant | |
wheat germ expression system |