Learn More
Abnova™ Human TSPAN7 Partial ORF (NP_004606, 113 a.a. - 212 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_004606 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7102 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16126755
|
Abnova™
H00007102-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16116755
|
Abnova™
H00007102-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein and may have a role in the control of neurite outgrowth. It is known to complex with integrins. This gene is associated with X-linked mental retardation and neuropsychiatric diseases such as Huntington's chorea, fragile X syndrome and myotonic dystrophy. [provided by RefSeq]
Sequence: RHEIKDTFLRTYTDAMQTYNGNDERSRAVDHVQRSLSCCGVQNYTNWSTSPYFLEHGIPPSCCMNETDCNPQDLHNLTVAATKVNQKGCYDLVTSFMETNSpecifications
NP_004606 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
A15/CCG-B7/CD231/DXS1692E/MRX58/MXS1/TALLA-1/TM4SF2/TM4SF2b | |
TSPAN7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
7102 | |
TSPAN7 (Human) Recombinant Protein (Q01) | |
RHEIKDTFLRTYTDAMQTYNGNDERSRAVDHVQRSLSCCGVQNYTNWSTSPYFLEHGIPPSCCMNETDCNPQDLHNLTVAATKVNQKGCYDLVTSFMETN | |
RUO | |
TSPAN7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |