missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TSC1 Partial ORF (NP_000359, 166 a.a. - 274 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_000359 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7248 |
Molecular Weight (g/mol) | 37.73kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16156995
|
Abnova™
H00007248-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16146995
|
Abnova™
H00007248-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a growth inhibitory protein thought to play a role in the stabilization of tuberin. Mutations in this gene have been associated with tuberous sclerosis. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: LKKPGHVAEVYLVHLHASVYALFHRLYGMYPCNFVSFLRSHYSMKENLETFEEVVKPMMEHVRIHPELVTGSKDHELDPRRWKRLETHDVVIECAKISLDPTEASYEDGSpecifications
NP_000359 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0243/LAM/MGC86987/TSC | |
TSC1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
7248 | |
TSC1 (Human) Recombinant Protein (Q01) | |
LKKPGHVAEVYLVHLHASVYALFHRLYGMYPCNFVSFLRSHYSMKENLETFEEVVKPMMEHVRIHPELVTGSKDHELDPRRWKRLETHDVVIECAKISLDPTEASYEDG | |
RUO | |
TSC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |