Learn More
Abnova™ Human TRIM63 Full-length ORF (NP_115977.2, 1 a.a. - 353 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_115977.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 84676 |
Molecular Weight (g/mol) | 66.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16106013
|
Abnova™
H00084676-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16196003
|
Abnova™
H00084676-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the RING zinc finger protein family found in striated muscle and iris. The product of this gene is localized to the Z-line and M-line lattices of myofibrils, where titin's N-terminal and C-terminal regions respectively bind to the sarcomere. In vitro binding studies have shown that this protein also binds directly to titin near the region of titin containing kinase activity. Another member of this protein family binds to microtubules. Since these family members can form heterodimers, this suggests that these proteins may serve as a link between titin kinase and microtubule-dependent signal pathways in muscle. [provided by RefSeq]
Sequence: MDYKSSLIQDGNPMENLEKQLICPICLEMFTKPVVILPCQHNLCRKCANDIFQAANPYWTSRGSSVSMSGGRFRCPTCRHEVIMDRHGVYGLQRNLLVENIIDIYKQECSSRPLQKGSHPMCKEHEDEKINIYCLTCEVPTCSMCKVFGIHKACEVAPLQSVFQGQKTELNNCISMLVAGNDRVQTIITQLEDSRRVTKENSHQVKEELSQKFDTLYAILDEKKSELLQRITQEQEKKLSFIEALIQQYQEQLDKSTKLVETAIQSLDEPGGATFLLTAKQLIKSIVEASKGCQLGKTEQGFENMDFFTLDLEHIADALRAIDFGTDEEEEEFIEEEDQEEEESTEGKEEGHQSpecifications
NP_115977.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MDYKSSLIQDGNPMENLEKQLICPICLEMFTKPVVILPCQHNLCRKCANDIFQAANPYWTSRGSSVSMSGGRFRCPTCRHEVIMDRHGVYGLQRNLLVENIIDIYKQECSSRPLQKGSHPMCKEHEDEKINIYCLTCEVPTCSMCKVFGIHKACEVAPLQSVFQGQKTELNNCISMLVAGNDRVQTIITQLEDSRRVTKENSHQVKEELSQKFDTLYAILDEKKSELLQRITQEQEKKLSFIEALIQQYQEQLDKSTKLVETAIQSLDEPGGATFLLTAKQLIKSIVEASKGCQLGKTEQGFENMDFFTLDLEHIADALRAIDFGTDEEEEEFIEEEDQEEEESTEGKEEGHQ | |
RUO | |
TRIM63 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
84676 | |
TRIM63 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ32380/IRF/MURF1/MURF2/RNF28/SMRZ | |
TRIM63 | |
Recombinant | |
wheat germ expression system |