Learn More
Abnova™ Human TRIM32 Partial ORF (NP_036342, 105 a.a. - 204 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00022954-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. The protein has also been localized to the nucleus, where it interacts with the activation domain of the HIV-1 Tat protein. The Tat protein activates transcription of HIV-1 genes. [provided by RefSeq]
Sequence: RRLPRQFCRSCGLVLCEPCREADHQPPGHCTLPVKEAAEERRRDFGEKLTRLRELMGELQRRKAALEGVSKDLQARYKAVLQEYGHEERRVQDELARSRKSpecifications
NP_036342 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RRLPRQFCRSCGLVLCEPCREADHQPPGHCTLPVKEAAEERRRDFGEKLTRLRELMGELQRRKAALEGVSKDLQARYKAVLQEYGHEERRVQDELARSRK | |
RUO | |
TRIM32 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
22954 | |
TRIM32 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BBS11/HT2A/LGMD2H/TATIP | |
TRIM32 | |
Recombinant | |
wheat germ expression system |