missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TRFP Full-length ORF (NP_004266.2, 1 a.a. - 212 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_004266.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9477 |
Molecular Weight (g/mol) | 49.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16173722
|
Abnova™
H00009477-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16163722
|
Abnova™
H00009477-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
MED20 is a subunit of the Mediator complex, a multiprotein coactivator of RNA transcription that interacts with DNA-bound transcriptional activators, RNA polymerase II (see MIM 180660), and general initiation factors (Sato et al., 2003 [PubMed 14576168]).[supplied by OMIM]
Sequence: MGVTCVSQMPVAEGKSVQQTVELLTRKLEMLGAEKQGTFCVDCETYHTAASTLGSQGQTGKLMYVMHNSEYPLSCFALFENGPCLIADTNFDVLMVKLKGFFQSAKASKIETRGTRYQYCDFLVKVGTVTMGPSARGISVEVEYGPCVVASDCWSLLLEFLQSFLGSHTPGAPAVFGNRHDAVYGPADTMVQYMELFNKIRKQQQVPVAGIRSpecifications
NP_004266.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGVTCVSQMPVAEGKSVQQTVELLTRKLEMLGAEKQGTFCVDCETYHTAASTLGSQGQTGKLMYVMHNSEYPLSCFALFENGPCLIADTNFDVLMVKLKGFFQSAKASKIETRGTRYQYCDFLVKVGTVTMGPSARGISVEVEYGPCVVASDCWSLLLEFLQSFLGSHTPGAPAVFGNRHDAVYGPADTMVQYMELFNKIRKQQQVPVAGIR | |
RUO | |
MED20 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9477 | |
TRFP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586D2223/MGC29869/PRO0213/TRFP | |
MED20 | |
Recombinant | |
wheat germ expression system |