missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TOR1A Partial ORF (AAH00674.1, 1 a.a. - 332 a.a. (del303E) ) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH00674.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1861 |
Molecular Weight (g/mol) | 62.04kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16107414
|
Abnova™
H00001861-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16117414
|
Abnova™
H00001861-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is a member of the AAA family of adenosine triphosphatases (ATPases), is related to the Clp protease/heat shock family and is expressed prominently in the substantia nigra pars compacta. Mutations in this gene result in the autosomal dominant disorder, torsion dystonia 1. [provided by RefSeq]
Sequence: MKLGRAVLGLLLLAPSVVQAVEPISLGLALAGVLTGYIYPRLYCLFAECCGQKRSLSREALQKDLDDNLFGQHLAKKIILNAVFGFINNPKPKKPLTLSLHGWTGTGKNFVSKIIAENIYEGGLNSDYVHLFVATLHFPHASNITLYKDQLQLWIRGNVSACARSIFIFDEMDKMHAGLIDAIKPFLDYYDLVDGVSYQKAMFIFLSNAGAERITDVALDFWRSGKQREDIKLKDIEHALSVSVFNNKNSGFWHSSLIHRNLIDYFVPFLPLEYKHLKMCIRVEMQSRGYEIDEDIVSRVAEMTFFPKEERVFSDKGCKTVFTKLDYYYDDSpecifications
AAH00674.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.04kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DQ2/DYT1 | |
TOR1A | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
1861 | |
TOR1A (Human) Recombinant Protein (Q01) | |
MKLGRAVLGLLLLAPSVVQAVEPISLGLALAGVLTGYIYPRLYCLFAECCGQKRSLSREALQKDLDDNLFGQHLAKKIILNAVFGFINNPKPKKPLTLSLHGWTGTGKNFVSKIIAENIYEGGLNSDYVHLFVATLHFPHASNITLYKDQLQLWIRGNVSACARSIFIFDEMDKMHAGLIDAIKPFLDYYDLVDGVSYQKAMFIFLSNAGAERITDVALDFWRSGKQREDIKLKDIEHALSVSVFNNKNSGFWHSSLIHRNLIDYFVPFLPLEYKHLKMCIRVEMQSRGYEIDEDIVSRVAEMTFFPKEERVFSDKGCKTVFTKLDYYYDD | |
RUO | |
TOR1A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |