Learn More
Abnova™ Human TLL1 Partial ORF (NP_036596.3, 77 a.a. - 150 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007092-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes an astacin-like zinc-dependent metalloprotease and is a subfamily member of the metzincin family. A similar protein in mice is required during heart development and specifically processes procollagen C-propeptides and chordin at similar cleavage sites. [provided by RefSeq]
Sequence: RTIDLTQNPFGNLGHTTGGLGDHAMSKKRGALYQLIDRIRRIGFGLEQNNTVKGKVPLQFSGQNEKNRVPRAATSpecifications
NP_036596.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.88kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RTIDLTQNPFGNLGHTTGGLGDHAMSKKRGALYQLIDRIRRIGFGLEQNNTVKGKVPLQFSGQNEKNRVPRAAT | |
RUO | |
TLL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7092 | |
TLL1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TLL | |
TLL1 | |
Recombinant | |
wheat germ expression system |