Learn More
ACROBiosystems Human TIMP1 Protein, His Tag
453.47€ - 2591.26€
Specifications
Accession Number | AAH00866 |
---|---|
Molecular Weight (g/mol) | 21.5 kDa |
Source | Human TIMP1, His Tag (TI1-H5226) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Ala 207 (Accession # P01033-1). |
Immunogen | Cys 24 - Ala 207 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15897757
|
ACROBiosystems
TI1-H5226-1MG |
1 mg |
2591.26€
1mg |
Estimated Shipment: 11-04-2024
Log in to see stock. |
|||||
15807767
|
ACROBiosystems
TI1-H5226-50UG |
50 μg |
453.47€
50µg |
Estimated Shipment: 11-04-2024
Log in to see stock. |
|||||
Description
TIMP metallopeptidase inhibitor 1 is also known as TIMP1, a tissue inhibitor of metalloproteinases, is a glycoprotein that is expressed from the several tissues of organisms. This protein a member of the TIMP family.There are four members of the family, TIMP1,TIMP2,TIMP3 and TIMP4. TIMP1 is a glycoprotein with a molecular mass of 28 kDa produced by a wide range of cell types. The glycoprotein is a natural inhibitor of the matrix metalloproteinases (MMPs), a group of peptidases involved in degradation of the extracellular matrix. In addition to its inhibitory role against most of the known MMPs, the encoded protein is able to promote cell proliferation in a wide range of cell types, and may also have an anti-apoptotic function. Transcription of this gene is highly inducible in response to many cytokines and hormones. In addition, the expression from some but not all inactive X chromosomes suggests that this gene inactivation is polymorphic in human females. This gene is located within intron 6 of the synapsin I gene and is transcribed in the opposite direction.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.5 kDa. The protein migrates as 30-34 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
AAH00866 | |
Human TIMP1, His Tag (TI1-H5226) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Ala 207 (Accession # P01033-1). | |
-20°C | |
TIMP-1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
21.5 kDa | |
Cys 24 - Ala 207 | |
TIMP1,CLGI,TIMP | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |