missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human THYN1 Full-length ORF (NP_954994.1, 1 a.a. - 166 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00029087-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16158982

  • 545.00€ / 25µg

Estimated Shipment: 24-08-2022
to see stock.
Add to basket

Description

Description

This gene encodes a protein that is highly conserved among vertebrates and plant species and may be involved in the induction of apoptosis. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]

Sequence: MSRPRKRLAGTSGSDKGLSGKRTKTENSGEALAKVEDSNPQKTSATKNCLKNLSSHWLMKSEPESRLEKGVDVKFSIEDLKAQPKQTTCWDGVRNYQARNFLRAMKLGEEAFFYHSNCKEPGIAGLMKIVKEAYPDHTQFEKNNPHYDPSSKEDNPKWSMKSLILF
Specifications

Specifications

NP_954994.1
Liquid
29087
THYN1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSRPRKRLAGTSGSDKGLSGKRTKTENSGEALAKVEDSNPQKTSATKNCLKNLSSHWLMKSEPESRLEKGVDVKFSIEDLKAQPKQTTCWDGVRNYQARNFLRAMKLGEEAFFYHSNCKEPGIAGLMKIVKEAYPDHTQFEKNNPHYDPSSKEDNPKWSMKSLILF
RUO
THYN1
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
45.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSPC144/MDS012/MGC12187/MY105/THY28/THY28KD
THYN1
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers