missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human THRSP Full-length ORF (NP_003242.1, 1 a.a. - 146 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007069-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16123638

  • 335.00€ / 10µg
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is similar to the gene product of S14, a rat gene whose expression is limited to liver and adipose tissue and is controlled by nutritional and hormonal factors. This gene has been shown to be expressed in liver and adipocytes, particularly in lipomatous modules. It is also found to be expressed in lipogenic breast cancers, which suggests a role in controlling tumor lipid metabolism. [provided by RefSeq]

Sequence: MQVLTKRYPKNCLLTVMDRYAAEVHNMEQVVMIPSLLRDVQLSGPGGQAQAEAPDLYTYFTMLKAICVDVDHGLLPREEWQAKVAGSEENGTAETEEVEDESASGELDLEAQFHLHFSSLHHILMHLTEKAQEVTRKYQEMTGQVW
Specifications

Specifications

NP_003242.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
43kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC21659/S14/SPOT14
THRSP
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
7069
THRSP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQVLTKRYPKNCLLTVMDRYAAEVHNMEQVVMIPSLLRDVQLSGPGGQAQAEAPDLYTYFTMLKAICVDVDHGLLPREEWQAKVAGSEENGTAETEEVEDESASGELDLEAQFHLHFSSLHHILMHLTEKAQEVTRKYQEMTGQVW
RUO
THRSP
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers