Learn More
Abnova™ Human TEF Full-length ORF (NP_003207.1, 1 a.a. - 303 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_003207.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7008 |
Molecular Weight (g/mol) | 59.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16116455
|
Abnova™
H00007008-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16126455
|
Abnova™
H00007008-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
Thyrotroph embryonic factor (TEF), a transcription factor, is a member of the PAR (proline and acidic amino acid-rich) subfamily of basic region/leucine zipper (bZIP) transcription factors. It is expressed in a broad range of cells and tissues in adult animals, however, during embryonic development, TEF expression appears to be restricted to the developing anterior pituitary gland, coincident with the appearance of thyroid-stimulating hormone, beta (TSHB). Indeed, TEF can bind to, and transactivate the TSHB promoter. It shows homology (in the functional domains) with other members of the PAR-bZIP subfamily of transcription factors, which include albumin D box-binding protein (DBP), human hepatic leukemia factor (HLF) and chicken vitellogenin gene-binding protein (VBP); VBP is considered the chicken homologue of TEF. Different members of the subfamily can readily form heterodimers, and share DNA-binding, and transcriptional regulatory properties. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MSDAGGGKKPPVDPQAGPGPGPGRAAGERGLSGSFPLVLKKLMENPPREARLDKEKGKEKLEEDEAAAASTMAVSASLMPPIWDKTIPYDGESFHLEYMDLDEFLLENGIPASPTHLAHNLLLPVAELEGKESASSSTASPPSSSTAIFQPSETVSSTESSLEKERETPSPIDPNCVEVDVNFNPDPADLVLSSVPGGELFNPRKHKFAEEDLKPQPMIKKAKKVFVPDEQKDEKYWTRRKKNNVAAKRSRDARRLKENQITIRAAFLEKENTALRTEVAELRKEVGKCKTIVSKYETKYGPLSpecifications
NP_003207.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSDAGGGKKPPVDPQAGPGPGPGRAAGERGLSGSFPLVLKKLMENPPREARLDKEKGKEKLEEDEAAAASTMAVSASLMPPIWDKTIPYDGESFHLEYMDLDEFLLENGIPASPTHLAHNLLLPVAELEGKESASSSTASPPSSSTAIFQPSETVSSTESSLEKERETPSPIDPNCVEVDVNFNPDPADLVLSSVPGGELFNPRKHKFAEEDLKPQPMIKKAKKVFVPDEQKDEKYWTRRKKNNVAAKRSRDARRLKENQITIRAAFLEKENTALRTEVAELRKEVGKCKTIVSKYETKYGPL | |
RUO | |
TEF | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7008 | |
TEF (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TEF | |
Wheat Germ (in vitro) | |
GST | |
Liquid |