missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TCL1A Partial ORF (AAH03574.1, 7 a.a. - 102 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008115-Q02.10ug
Additional Details : Weight : 0.02000kg
Description
Sequence: LGEAVTDHPDRLWAWEKFVYLDEKQHAWLPLTIEIKDRLQLRVLLRREDVVLGRPMTPTQIGPSLLPIMWQLYPDGRYRSSDSSFWRLVYHIKIDGSpecifications
AAH03574.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.286kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LGEAVTDHPDRLWAWEKFVYLDEKQHAWLPLTIEIKDRLQLRVLLRREDVVLGRPMTPTQIGPSLLPIMWQLYPDGRYRSSDSSFWRLVYHIKIDG | |
RUO | |
TCL1A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8115 | |
TCL1A (Human) Recombinant Protein (Q02) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TCL1 | |
TCL1A | |
Recombinant | |
wheat germ expression system |