Learn More
Abnova™ Human SYN3 Full-length ORF (AAH75065.1, 1 a.a. - 580 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH75065.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8224 |
Molecular Weight (g/mol) | 89.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16112322
|
Abnova™
H00008224-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
16102322
|
Abnova™
H00008224-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
This gene is a member of the synapsin gene family. Synapsins encode neuronal phosphoproteins which associate with the cytoplasmic surface of synaptic vesicles. Family members are characterized by common protein domains, and they are implicated in synaptogenesis and the modulation of neurotransmitter release, suggesting a potential role in several neuropsychiatric diseases. The protein encoded by this gene shares the synapsin family domain model, with domains A, C, and E exhibiting the highest degree of conservation. The protein contains a unique domain J, located between domains C and E. Based on this gene's localization to 22q12.3, a possible schizophrenia susceptibility locus, and the established neurobiological roles of the synapsins, this family member may represent a candidate gene for schizophrenia. The TIMP3 gene is located within an intron of this gene and is transcribed in the opposite direction. Alternative splicing of this gene results in multiple splice variants that encode different isoforms. [provided by RefSeq]
Sequence: MNFLRRRLSDSSFMANLPNGYMTDLQRPDSSTSSPASPAMERRHPQPLAASFSSPGSSLFSSLSSAMKQAPQATSGLMEPPGPSTPIVQRPRILLVIDDAHTDWSKYFHGKKVNGEIEIRVEQAEFSELNLAAYVTGGCMVDMQVVRNGTKVVSRSFKPDFILVRQHAYSMALGEDYRSLVIGLQYGGLPAVNSLYSVYNFCSKPWVFSQLIKIFHSLGPEKFPLVEQTFFPNHKPMVTAPHFPVVVKLGHAHAGMGKIKVENQLDFQDITSVVAMAKTYATTEAFIDSKYDIRIQKIGSNYKAYMRTSISGNWKANTGSAMLEQVAMTERYRLWVDSCSEMFGGLDICAVKAVHSKDGRDYIIEVMDSSMPLIGEHVEEDRQLMADLVVSKMSQLPMPGGTAPSPLRPWAPQIKSAKSPGQAQLGPQLGQPQPRPPPQGGPRQAQSPQPQRSGSPSQQRLSPQGQQPLSPQSGSPQQQRSPGSPQLSRASSGSSPNQASKPGATLASQPRPPVQGRSTSQQGEESKKPAPPHPHLNKSQSLTNSLSTSDTSQRGTPSEDEAKAETIRNLRKSFASLFSDSpecifications
AAH75065.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
89.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MNFLRRRLSDSSFMANLPNGYMTDLQRPDSSTSSPASPAMERRHPQPLAASFSSPGSSLFSSLSSAMKQAPQATSGLMEPPGPSTPIVQRPRILLVIDDAHTDWSKYFHGKKVNGEIEIRVEQAEFSELNLAAYVTGGCMVDMQVVRNGTKVVSRSFKPDFILVRQHAYSMALGEDYRSLVIGLQYGGLPAVNSLYSVYNFCSKPWVFSQLIKIFHSLGPEKFPLVEQTFFPNHKPMVTAPHFPVVVKLGHAHAGMGKIKVENQLDFQDITSVVAMAKTYATTEAFIDSKYDIRIQKIGSNYKAYMRTSISGNWKANTGSAMLEQVAMTERYRLWVDSCSEMFGGLDICAVKAVHSKDGRDYIIEVMDSSMPLIGEHVEEDRQLMADLVVSKMSQLPMPGGTAPSPLRPWAPQIKSAKSPGQAQLGPQLGQPQPRPPPQGGPRQAQSPQPQRSGSPSQQRLSPQGQQPLSPQSGSPQQQRSPGSPQLSRASSGSSPNQASKPGATLASQPRPPVQGRSTSQQGEESKKPAPPHPHLNKSQSLTNSLSTSDTSQRGTPSEDEAKAETIRNLRKSFASLFSD | |
RUO | |
SYN3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8224 | |
SYN3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SYN3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |