missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SUPT4H1 Full-length ORF (AAH02802, 1 a.a. - 117 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006827-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121152

  • 335.00€ / 10µg
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

Sequence: MALETVPKDLRHLRACLLCSLVKTIDQFEYDGCDNCDAYLQMKGNREMVYDCTSSSFDGIIAMMSPEDSWVSKWQRVSNFKPGVYAVSVTGRLPQGIVRELKSRGVAYKSRDTAIKT
Specifications

Specifications

AAH02802
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.61kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SPT4/SPT4H/SUPT4H
SUPT4H1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
6827
SUPT4H1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MALETVPKDLRHLRACLLCSLVKTIDQFEYDGCDNCDAYLQMKGNREMVYDCTSSSFDGIIAMMSPEDSWVSKWQRVSNFKPGVYAVSVTGRLPQGIVRELKSRGVAYKSRDTAIKT
RUO
SUPT4H1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers