missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human STAM2 Full-length ORF (AAH28740, 1 a.a. - 525 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH28740 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10254 |
Molecular Weight (g/mol) | 83.49kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16124742
|
Abnova™
H00010254-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16114742
|
Abnova™
H00010254-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is closely related to STAM, an adaptor protein involved in the downstream signaling of cytokine receptors, both of which contain a SH3 domain and the immunoreceptor tyrosine-based activation motif (ITAM). Similar to STAM, this protein acts downstream of JAK kinases, and is phosphorylated in response to cytokine stimulation. This protein and STAM thus are thought to exhibit compensatory effects on the signaling pathway downstream of JAK kinases upon cytokine stimulation. [provided by RefSeq]
Sequence: MPLFTANPFEQDVEKATNEYNTTEDWSLIMDICDKVGSTPNGAKDCLKAIMKRVNHKVPHVALQALTLLGACVANCGKIFHLEVCSRDFATEVRAVIKNKAHPKVCEKLKSLMVEWSEEFQKDPQFSLISATIKSMKEEGITFPPAGSQTVSAAAKNGTSSNKNKEDEDIAKAIELSLQEQKQQHTETKSLYPSSEIQLNNKVARKVRALYDFEAVEDNELTFKHGEIIIVLDDSDANWWKGENHRGIGLFPSNFVTTNLNIETEAAAVDKLNVIDDDVEEIKKSEPEPVYIDEDKMDRALQVLQSIDPTDSKPDSQDLLDLEDICQQMGPMIDEKLEEIDRKHSELSELNVKVLEALELYNKLVNEAPVYSVYSKLHPPAHYPPASSGVPMQTYPVQSHGGNYMGQSIHQVTVAQSYSLGPDQIGPLRSLPPNVNSSVTAQPAQTSYLSTGQDTVSNPTYMNQNSNLQSATGTTAYTQQMGMSVDMSSYQNTTSNLPQLAGFPVTVPAHPVAQQHTNYHQQPLLSpecifications
AAH28740 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
83.49kDa | |
Glutathione Sepharose 4 Fast Flow | |
MPLFTANPFEQDVEKATNEYNTTEDWSLIMDICDKVGSTPNGAKDCLKAIMKRVNHKVPHVALQALTLLGACVANCGKIFHLEVCSRDFATEVRAVIKNKAHPKVCEKLKSLMVEWSEEFQKDPQFSLISATIKSMKEEGITFPPAGSQTVSAAAKNGTSSNKNKEDEDIAKAIELSLQEQKQQHTETKSLYPSSEIQLNNKVARKVRALYDFEAVEDNELTFKHGEIIIVLDDSDANWWKGENHRGIGLFPSNFVTTNLNIETEAAAVDKLNVIDDDVEEIKKSEPEPVYIDEDKMDRALQVLQSIDPTDSKPDSQDLLDLEDICQQMGPMIDEKLEEIDRKHSELSELNVKVLEALELYNKLVNEAPVYSVYSKLHPPAHYPPASSGVPMQTYPVQSHGGNYMGQSIHQVTVAQSYSLGPDQIGPLRSLPPNVNSSVTAQPAQTSYLSTGQDTVSNPTYMNQNSNLQSATGTTAYTQQMGMSVDMSSYQNTTSNLPQLAGFPVTVPAHPVAQQHTNYHQQPLL | |
RUO | |
STAM2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10254 | |
STAM2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp564C047/Hbp/STAM2A/STAM2B | |
STAM2 | |
Recombinant | |
wheat germ expression system |