missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SPPL2A Partial ORF (NP_116191, 31 a.a. - 129 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifications

Accession Number NP_116191
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 84888
Molecular Weight (g/mol) 36.63kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16160927
View Documents Promotion Details
Abnova™
H00084888-Q01.25UG
25 ug
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
 
16150927
View Documents Promotion Details
Abnova™
H00084888-Q01.10UG
10 ug
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
 
Description

Description

This gene is a member of the signal peptide peptidase-like protease (SPPL) family and encodes an endosomal membrane protein with a protease associated (PA) domain. This protein plays a role in innate and adaptive immunity. A pseudogene of this gene also lies on chromosome 15. [provided by RefSeq]

Sequence: HASGNGTTKDYCMLYNPYWTALPSTLENATSISLMNLTSTPLCNLSDIPPVGIKSKAVVVPWGSCHFLEKARIAQKGGAEAMLVVNNSVLFPPSGNRSE
Specifications

Specifications

NP_116191
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IMP3/PSL2
SPPL2A
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
84888
SPPL2A (Human) Recombinant Protein (Q01)
HASGNGTTKDYCMLYNPYWTALPSTLENATSISLMNLTSTPLCNLSDIPPVGIKSKAVVVPWGSCHFLEKARIAQKGGAEAMLVVNNSVLFPPSGNRSE
RUO
SPPL2A
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications