Learn More
Abnova™ Human SOX5 Partial ORF (NP_008871.3, 181 a.a. - 230 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006660-Q01.25ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The encoded protein may play a role in chondrogenesis. A pseudogene of this gene is located on chromosome 8. Multiple transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: GSGNFGEIKGTPESLAEKERQLMGMINQLTSLREQLLAAHDEQKKLAASQSpecifications
NP_008871.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.24kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GSGNFGEIKGTPESLAEKERQLMGMINQLTSLREQLLAAHDEQKKLAASQ | |
RUO | |
SOX5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6660 | |
SOX5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
L-SOX5/MGC35153 | |
SOX5 | |
Recombinant | |
wheat germ expression system |