missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SOAT1 Partial ORF (NP_003092.4, 249 a.a. - 321 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_003092.4 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6646 |
Molecular Weight (g/mol) | 33.77kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16094735
|
Abnova™
H00006646-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16084735
|
Abnova™
H00006646-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
Acyl-coenzyme A:cholesterol acyltransferase (ACACT; EC 2.3.1.26) is an intracellular protein located in the endoplasmic reticulum that forms cholesterol esters from cholesterol. Accumulation of cholesterol esters as cytoplasmic lipid droplets within macrophages and smooth muscle cells is a characteristic feature of the early stages of atherosclerotic plaques (Cadigan et al., 1988 [PubMed 3335499]).[supplied by OMIM]
Sequence: PPASRFIIIFEQIRFVMKAHSFVRENVPRVLNSAKEKSSTVPIPTVNQYLYFLFAPTLIYRDSYPRNPTVRWGSpecifications
NP_003092.4 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.77kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACACT/ACAT/ACAT1/RP11-215I23.2/SOAT/STAT | |
SOAT1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
6646 | |
SOAT1 (Human) Recombinant Protein (Q01) | |
PPASRFIIIFEQIRFVMKAHSFVRENVPRVLNSAKEKSSTVPIPTVNQYLYFLFAPTLIYRDSYPRNPTVRWG | |
RUO | |
SOAT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |