Learn More
Abnova™ Human SNX6 Full-length ORF (AAH01798, 1 a.a. - 290 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH01798 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 58533 |
Molecular Weight (g/mol) | 57.64kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16163123
|
Abnova™
H00058533-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16173123
|
Abnova™
H00058533-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein associates with the long isoform of the leptin receptor, the transforming growth factor-beta family of receptor serine-threonine kinases, and with receptor tyrosine kinases for platelet-derived growth factor, insulin, and epidermal growth factor. This protein may form oligomeric complexes with family member proteins through interactions of both the PX domain and the coiled coil regions of the molecules. Translocation of this protein from the cytoplasm to the nucleus occurs after binding to proviral integration site 1 protein. This gene results in two transcripts encoding two distinct isoforms. [provided by RefSeq]
Sequence: MTKEEFTKMKQELEAEYLAIFKKTVAMHEVFLCRVAAHPILGRDLNFHVFLEYNQDLSVRGKNKKEKLEDFFKNMVKSADGVIVSGVKDVDDFFEHERTFLLEYHNRVKDASAKSDRMTRSHKSAADDYNRIGSSLYALGTQDSTDICKFFLKVSELFDKTRKIEARVSADEDLKLSDLLKYYLRESQAAKDLLYRRSRSLVDYENANKALDKARAKNKDVLQAETSQQLCCQKFEKISESAKQELIDFKTRRVAAFRKNLVELAELELKHAKGNLQLLQNCLAVLNGDTSpecifications
AAH01798 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
57.64kDa | |
Glutathione Sepharose 4 Fast Flow | |
MTKEEFTKMKQELEAEYLAIFKKTVAMHEVFLCRVAAHPILGRDLNFHVFLEYNQDLSVRGKNKKEKLEDFFKNMVKSADGVIVSGVKDVDDFFEHERTFLLEYHNRVKDASAKSDRMTRSHKSAADDYNRIGSSLYALGTQDSTDICKFFLKVSELFDKTRKIEARVSADEDLKLSDLLKYYLRESQAAKDLLYRRSRSLVDYENANKALDKARAKNKDVLQAETSQQLCCQKFEKISESAKQELIDFKTRRVAAFRKNLVELAELELKHAKGNLQLLQNCLAVLNGDT | |
RUO | |
SNX6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
58533 | |
SNX6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC3157/MSTP010/TFAF2 | |
SNX6 | |
Recombinant | |
wheat germ expression system |