missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SMAD6 Partial ORF (NP_005576, 285 a.a. - 384 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004091-Q01.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16024365

  • 518.16€ / 25µg

Estimated Shipment: 21-01-2022
Add to basket

Description

Description

The protein encoded by this gene belongs to the SMAD family of proteins, which are related to Drosophila 'mothers against decapentaplegic' (Mad) and C. elegans Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein functions in the negative regulation of BMP and TGF-beta/activin-signalling. Multiple transcript variants encoding different isoforms have been found for this gene

Sequence: RDEYKPLDLSDSTLSYTETEATNSLITAPGEFSDASMSPDATKPSHWCSVAYWEHRTRVGRLYAVYDQAVSIFYDLPQGSGFCLGQLNLEQRSESVRRTR
Specifications

Specifications

NP_005576
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue
RDEYKPLDLSDSTLSYTETEATNSLITAPGEFSDASMSPDATKPSHWCSVAYWEHRTRVGRLYAVYDQAVSIFYDLPQGSGFCLGQLNLEQRSESVRRTR
RUO
SMAD6
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
4091
SMAD6 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HsT17432/MADH6/MADH7
SMAD6
Yes
Videos
SDS
Documents
Certificates
Special Offers

Special Offers