missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC9A6 Partial ORF (NP_006350, 602 a.a. - 669 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_006350 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10479 |
Molecular Weight (g/mol) | 33.22kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16102626
|
Abnova™
H00010479-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
16112626
|
Abnova™
H00010479-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
The SLC9A6 gene encodes a monovalent sodium-selective sodium/hydrogen exchanger (NHE) that is found in the membranes of intracellular organelles such as mitochondria and endosomes. NHEs participate in a wide array of essential cellular processes, including control of intracellular pH, maintenance of cellular volume, and reabsorption of sodium across renal, intestinal, and other epithelia.[supplied by OMIM]
Sequence: YGDSTVNTEPATSSAPRRFMGNSSEDALDRELAFGDHELVIRGTRLVLPMDDSEPPLNLLDNTRHGPASpecifications
NP_006350 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.22kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0267/MRSA/NHE6 | |
SLC9A6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10479 | |
SLC9A6 (Human) Recombinant Protein (Q01) | |
YGDSTVNTEPATSSAPRRFMGNSSEDALDRELAFGDHELVIRGTRLVLPMDDSEPPLNLLDNTRHGPA | |
RUO | |
SLC9A6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |