missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC3A2 Partial ORF (NP_002385.3, 203 a.a. - 312 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifications

Accession Number NP_002385.3
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6520
Molecular Weight (g/mol) 37.84kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16135385
View Documents Promotion Details
Abnova™
H00006520-Q01.25UG
25 ug
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
16125385
View Documents Promotion Details
Abnova™
H00006520-Q01.10UG
10 ug
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

This gene is a member of the solute carrier family and encodes a cell surface, transmembrane protein with an alpha amylase domain. The protein exists as the heavy chain of a heterodimer, covalently bound through di-sulfide bonds to one of several possible light chains. It associates with integrins and mediates integrin-dependent signaling related to normal cell growth and tumorigenesis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]

Sequence: IIVRAPRCRELPAQKWWHTGALYRIGDLQAFQGHGAGNLAGLKGRLDYLSSLKVKGLVLGPIHKNQKDDVAQTDLLQIDPNFGSKEDFDSLLQSAKKKSIRVILDLTPNY
Specifications

Specifications

NP_002385.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
4F2/4F2HC/4T2HC/CD98/CD98HC/MDU1/NACAE
SLC3A2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
6520
SLC3A2 (Human) Recombinant Protein (Q01)
IIVRAPRCRELPAQKWWHTGALYRIGDLQAFQGHGAGNLAGLKGRLDYLSSLKVKGLVLGPIHKNQKDDVAQTDLLQIDPNFGSKEDFDSLLQSAKKKSIRVILDLTPNY
RUO
SLC3A2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications