missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC39A13 Partial ORF (NP_689477.2, 170 a.a. - 226 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_689477.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 91252 |
Molecular Weight (g/mol) | 32.01kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16111157
|
Abnova™
H00091252-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16101157
|
Abnova™
H00091252-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
Zinc is an essential cofactor for hundreds of enzymes. It is involved in protein, nucleic acid, carbohydrate, and lipid metabolism, as well as in the control of gene transcription, growth, development, and differentiation. SLC39A13 belongs to a subfamily of proteins that show structural characteristics of zinc transporters (Taylor and Nicholson, 2003 [PubMed 12659941]).[supplied by OMIM]
Sequence: LDSKEEGTSQAPNKDPTAAAAALNGGHCLAQPAAEPGLGAVVRSIKVSGYLNLLANTSpecifications
NP_689477.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.01kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ25785 | |
SLC39A13 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
91252 | |
SLC39A13 (Human) Recombinant Protein (Q01) | |
LDSKEEGTSQAPNKDPTAAAAALNGGHCLAQPAAEPGLGAVVRSIKVSGYLNLLANT | |
RUO | |
SLC39A13 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |