missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC35A2 Partial ORF (NP_005651, 88 a.a. - 156 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007355-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107265

  • 349.00€ / 10µg

Estimated Shipment: 07-07-2023
to see stock.
Add to basket

Description

Description

Sequence: AQKRGNVKHLVLFLHEAVLVQYVDTLKLAVPSLIYTLQNNLQYVAISNLPAATFQVTYQLKILTTALFS
Specifications

Specifications

NP_005651
Liquid
7355
SLC35A2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
UGALT/UGAT/UGT/UGT1/UGT2/UGTL
SLC35A2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.33kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AQKRGNVKHLVLFLHEAVLVQYVDTLKLAVPSLIYTLQNNLQYVAISNLPAATFQVTYQLKILTTALFS
RUO
SLC35A2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers