missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC26A9 Partial ORF (NP_443166, 496 a.a. - 605 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_443166 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 115019 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16101467
|
Abnova™
H00115019-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16111467
|
Abnova™
H00115019-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
This gene is one member of a family of sulfate/anion transporter genes. Family members are well conserved in their genomic (number and size of exons) and protein (aa length among species) structures yet have markedly different tissue expression patterns. The product of this gene is a highly selective chloride ion channel regulated by WNK kinases. Alternative splicing results in multiple transcript variants encoding differing isoforms
Sequence: QTQFRNGYALAQVMDTDIYVNPKTYNRAQDIQGIKIITYCSPLYFANSEIFRQKVIAKTGMDPQKVLLAKQKYLKKQEKRRMRPTQQRRSLFMKTKTVSLQELQQDFENASpecifications
NP_443166 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SLC26A9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
115019 | |
SLC26A9 (Human) Recombinant Protein (Q01) | |
QTQFRNGYALAQVMDTDIYVNPKTYNRAQDIQGIKIITYCSPLYFANSEIFRQKVIAKTGMDPQKVLLAKQKYLKKQEKRRMRPTQQRRSLFMKTKTVSLQELQQDFENA | |
RUO | |
SLC26A9 | |
Recombinant | |
wheat germ expression system |