missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC23A2 Full-length ORF (AAH13112.1, 1 a.a. - 425 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH13112.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9962 |
Molecular Weight (g/mol) | 72.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16114292
|
Abnova™
H00009962-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16104292
|
Abnova™
H00009962-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
The absorption of vitamin C into the body and its distribution to organs requires two sodium-dependent vitamin C transporters. This gene encodes one of the two required transporters and the encoded protein accounts for tissue-specific uptake of vitamin C. Previously, this gene had an official symbol of SLC23A1. [provided by RefSeq]
Sequence: MMGIGKNTTSKSMEAGSSTEGKYEDEAKHPAFFTLPVVINGGATSSGEQDNEDTELMAIYTTENGIAEKSSLAETLDSTGSLDPQRSDMIYTIEDVPPWYLCIFLGLQHYLTCFSGTIAVPFLLADAMCVGYDQWATSQLIGTIFFCVGITTLLQTTFGCRLPLFQASAFAFLAPARAILSLDKWKCNTTDVSVANGTAELLHTEHIWYPRIREIQGAIIMSSLIEVVIGLLGLPGALLKYIGPLTITPTVALIGLSGFQAAGERAGKHWGIAMLTIFLVLLFSQYARNVKFPLPIYKSKKGWTAYKLQLFKMFPIILAILVSWLLCFIFTVTDVFPPDSTKYGFYARTDARQGVLLVAPWFKVPYPFQWGLPTVSAAGVIGMLSAVVASIIESIGDYYACARLSCAPPPPIHAINRYVPEKTSSSpecifications
AAH13112.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MMGIGKNTTSKSMEAGSSTEGKYEDEAKHPAFFTLPVVINGGATSSGEQDNEDTELMAIYTTENGIAEKSSLAETLDSTGSLDPQRSDMIYTIEDVPPWYLCIFLGLQHYLTCFSGTIAVPFLLADAMCVGYDQWATSQLIGTIFFCVGITTLLQTTFGCRLPLFQASAFAFLAPARAILSLDKWKCNTTDVSVANGTAELLHTEHIWYPRIREIQGAIIMSSLIEVVIGLLGLPGALLKYIGPLTITPTVALIGLSGFQAAGERAGKHWGIAMLTIFLVLLFSQYARNVKFPLPIYKSKKGWTAYKLQLFKMFPIILAILVSWLLCFIFTVTDVFPPDSTKYGFYARTDARQGVLLVAPWFKVPYPFQWGLPTVSAAGVIGMLSAVVASIIESIGDYYACARLSCAPPPPIHAINRYVPEKTSS | |
RUO | |
SLC23A2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9962 | |
SLC23A2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0238/NBTL1/SLC23A1/SVCT2/YSPL2 | |
SLC23A2 | |
Recombinant | |
wheat germ expression system |