missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human SLAMF1 / SLAM / CD150 Protein, His Tag

Brand:  ACROBiosystems SL1-H52H0-100UG

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15817707

  • 262.28€ / 100µg


Estimated Shipment: 22-10-2021
Add to basket

Description

Description

Self-ligand receptor of the signaling lymphocytic activation molecule (SLAM) family, also known as CD150, is the prototypic member of the SLAM subfamily of the CD2 protein family.SLAM receptors triggered by homo- or heterotypic cell-cell interactions are modulating the activation and differentiation of a wide variety of immune cells and thus are involved in the regulation and interconnection of both innate and adaptive immune response. Activities are controlled by presence or absence of small cytoplasmic adapter proteins, SH2D1A/SAP and/or SH2D1B/EAT-2. SLAMF1-induced signal-transduction events in T-lymphocytes are different from those in B-cells. The type I transmembrane glycoprotein Signaling Lymphocytic Activation Molecule (SLAM).

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.2 kDa. The protein migrates as 42-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

NP_003028.1
HEK293
Human
26.2 kDa
100 µg
Ala 21 - Pro 237
SLAMF1,CD150,CDw150,SLAM,IPO-3
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins
Powder
>95% as determined by SDS-PAGE
Human SLAMF1, His Tag (SL1-H52H0) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Pro 237 (Accession # Q13291-1).
-20°C
SLAMF1
His Tag
Videos
SDS
Documents
Certificates