Learn More
Abnova™ Human SGKL Partial ORF (AAH15326, 1 a.a. - 130 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023678-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the Ser/Thr protein kinase family and encodes a phosphoprotein with a PX (phox homology) domain. The protein phosphorylates several target proteins and has a role in neutral amino acid transport and activation of potassium and chloride channels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: MQRDHTMDYKESCPSVSIPSSDEHREKKKRFTVYKVLVSVGRSEWFVFRRYAEFDKLYNTLKKQFPAMALKIPAKRIFGDNFDPDFIKQRRAGLNEFIQNLVRYPELYNHPDVRAFLQMDSPKHQSDPSESpecifications
AAH15326 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
39.93kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQRDHTMDYKESCPSVSIPSSDEHREKKKRFTVYKVLVSVGRSEWFVFRRYAEFDKLYNTLKKQFPAMALKIPAKRIFGDNFDPDFIKQRRAGLNEFIQNLVRYPELYNHPDVRAFLQMDSPKHQSDPSE | |
RUO | |
SGK3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23678 | |
SGKL (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CISK/DKFZp781N0293/SGK2/SGKL | |
SGK3 | |
Recombinant | |
wheat germ expression system |