Learn More
Abnova™ Human SETX Partial ORF (NP_055861.2, 2579 a.a. - 2676 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023064-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein named for its homology to the Sen1p protein of fungi which has RNA helicase activity encoded by a domain at the C-terminal end of the protein. The protein encoded by this gene contains a DNA/RNA helicase domain at its C-terminal end which suggests that it may be involved in both DNA and RNA processing. Mutations in this gene have been associated with ataxia-ocular apraxia-2 (AOA2) and an autosomal dominant form of juvenile amyotrophic lateral sclerosis (ALS4). [provided by RefSeq]
Sequence: VVHQDLSHIQQPAAVVAALSSHKPPVRGEPPAASPEASTCQSKCDDPEEELCHRREARAFSEGEQEKCGSETHHTRRNSRWDKRTLEQEDSSSKKRKLSpecifications
NP_055861.2 | |
Liquid | |
23064 | |
SETX (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALS4/AOA2/DKFZp781B151/FLJ12840/FLJ43459/KIAA0625/SCAR1/bA479K20.2 | |
SETX | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VVHQDLSHIQQPAAVVAALSSHKPPVRGEPPAASPEASTCQSKCDDPEEELCHRREARAFSEGEQEKCGSETHHTRRNSRWDKRTLEQEDSSSKKRKL | |
RUO | |
SETX | |
Wheat Germ (in vitro) | |
GST |