missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SCN8A Partial ORF (NP_055006, 1854 a.a. - 1951 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006334-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16124975

  • 335.00€ / 10µg
Estimated Shipment: 27-05-2024
to see stock.
Add to basket

Description

Description

Voltage-dependent sodium channels, such as SCN8A, are responsible for the initial membrane depolarization that occurs during generation of action potentials in most electrically excitable cells (Plummer et al., 1998 [PubMed 9828131]).[supplied by OMIM]

Sequence: RVLGDSGELDILRQQMEERFVASNPSKVSYEPITTTLRRKQEEVSAVVLQRAYRGHLARRGFICKKTTSNKLENGGTHREKKESTPSTASLPSYDSVT
Specifications

Specifications

NP_055006
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RVLGDSGELDILRQQMEERFVASNPSKVSYEPITTTLRRKQEEVSAVVLQRAYRGHLARRGFICKKTTSNKLENGGTHREKKESTPSTASLPSYDSVT
RUO
SCN8A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6334
SCN8A (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CerIII/MED/NaCh6/Nav1.6/PN4
SCN8A
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers