missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SCARB2 Partial ORF (NP_005497, 339 a.a. - 437 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_005497 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 950 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16102861
|
Abnova™
H00000950-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16112861
|
Abnova™
H00000950-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is a type III glycoprotein that is located primarily in limiting membranes of lysosomes and endosomes. Studies of the similar protein in mice and rat suggested that this protein may participate in membrane transportation and the reorganization of endosomal/lysosomal compartment. Deficiency of the similar protein in mice was reported to impair cell membrane transport processes and cause pelvic junction obstruction, deafness, and peripheral neuropathy. [provided by RefSeq]
Sequence: FPHFYQADERFVSAIEGMHPNQEDHETFVDINPLTGIILKAAKRFQINIYVKKLDDFVETGDIRTMVFPVMYLNESVHIDKETASRLKSMINTTLIITNSpecifications
NP_005497 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AMRF/CD36L2/HLGP85/LIMPII/SR-BII | |
SCARB2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
950 | |
SCARB2 (Human) Recombinant Protein (Q01) | |
FPHFYQADERFVSAIEGMHPNQEDHETFVDINPLTGIILKAAKRFQINIYVKKLDDFVETGDIRTMVFPVMYLNESVHIDKETASRLKSMINTTLIITN | |
RUO | |
SCARB2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |