Learn More
Abnova™ Human SC65 Full-length ORF (NP_006446.1, 1 a.a. - 437 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010609-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This nucleolar protein was first characterized because it was an autoantigen in cases on interstitial cystitis. The protein, with a predicted molecular weight of 50 kDa, appears to be localized in the particulate compartment of the interphase nucleolus, with a distribution distinct from that of nucleolar protein B23. During mitosis it is associated with chromosomes. [provided by RefSeq]
Sequence: MARVAWGLLWLLLGSAGAQYEKYSFRGFPPEDLMPLAAAYGHALEQYEGESWRESARYLEAALRLHRLLRDSEAFCHANCSGPAPAAKPDPDGGRADEWACELRLFGRVLERAACLRRCKRTLPAFQVPYPPRQLLRDFQSRLPYQYLHYALFKANRLEKAVAAAYTFLQRNPKHELTAKYLNYYQGMLDVADESLTDLEAQPYEAVFLRAVKLYNSGDFRSSTEDMERALSEYLAVFARCLAGCEGAHEQVDFKDFYPAIADLFAESLQCKVDCEANLTPNVGGYFVDKFVATMYHYLQFAYYKLNDVRQAARSAASYMLFDPKDSVMQQNLVYYRFHRARWGLEEEDFQPREEAMLYHNQTAELRELLEFTHMYLQSDDEMELEETEPPLEPEDALSDAEFEGEGDYEEGMYADWWQEPDAKGDEAEAEPEPELASpecifications
NP_006446.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NOL55 | |
SC65 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10609 | |
SC65 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MARVAWGLLWLLLGSAGAQYEKYSFRGFPPEDLMPLAAAYGHALEQYEGESWRESARYLEAALRLHRLLRDSEAFCHANCSGPAPAAKPDPDGGRADEWACELRLFGRVLERAACLRRCKRTLPAFQVPYPPRQLLRDFQSRLPYQYLHYALFKANRLEKAVAAAYTFLQRNPKHELTAKYLNYYQGMLDVADESLTDLEAQPYEAVFLRAVKLYNSGDFRSSTEDMERALSEYLAVFARCLAGCEGAHEQVDFKDFYPAIADLFAESLQCKVDCEANLTPNVGGYFVDKFVATMYHYLQFAYYKLNDVRQAARSAASYMLFDPKDSVMQQNLVYYRFHRARWGLEEEDFQPREEAMLYHNQTAELRELLEFTHMYLQSDDEMELEETEPPLEPEDALSDAEFEGEGDYEEGMYADWWQEPDAKGDEAEAEPEPELA | |
RUO | |
SC65 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |