Learn More
Abnova™ Human SART3 Full-length ORF (AAH41638.1, 1 a.a. - 129 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH41638.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9733 |
Molecular Weight (g/mol) | 40.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16124052
|
Abnova™
H00009733-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16114052
|
Abnova™
H00009733-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is an RNA-binding nuclear protein that is a tumor-rejection antigen. This antigen possesses tumor epitopes capable of inducing HLA-A24-restricted and tumor-specific cytotoxic T lymphocytes in cancer patients and may be useful for specific immunotherapy. This gene product is found to be an important cellular factor for HIV-1 gene expression and viral replication. It also associates transiently with U6 and U4/U6 snRNPs during the recycling phase of the spliceosome cycle. This encoded protein is thought to be involved in the regulation of mRNA splicing. [provided by RefSeq]
Sequence: MATAAETSASEPEAESKAGPKADGEEDEVKAARTRRKVLSRAVAAATYKTMGPAWDQQEEGVSESDGDEYAMASSAESSPGEYEWEYDEEEEKNQLEIERLEEQVGPGVGSGHLPVFQVLGSPCPGPPPSpecifications
AAH41638.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
40.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MATAAETSASEPEAESKAGPKADGEEDEVKAARTRRKVLSRAVAAATYKTMGPAWDQQEEGVSESDGDEYAMASSAESSPGEYEWEYDEEEEKNQLEIERLEEQVGPGVGSGHLPVFQVLGSPCPGPPP | |
RUO | |
SART3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9733 | |
SART3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DSAP1/KIAA0156/MGC138188/P100/RP11-13G14/TIP110/p110/p110(nrb) | |
SART3 | |
Recombinant | |
wheat germ expression system |