missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RRAS Partial ORF (AAH16286, 109 a.a. - 218 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006237-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16164775

  • 545.00€ / 25µg

Estimated Shipment: 22-08-2022
to see stock.
Add to basket

Description

Description

Sequence: AINDRQSFNEVGKLFTQILRVKDRDDFPVVLVGNKADLESQRQVPRSEASAFGASHHVAYFEASAKLRLNVDEAFEQLVRAVRKYQEQELPPSPPSAPRKKGGGCPCVLL
Specifications

Specifications

AAH16286
Liquid
6237
RRAS (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RRAS
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AINDRQSFNEVGKLFTQILRVKDRDDFPVVLVGNKADLESQRQVPRSEASAFGASHHVAYFEASAKLRLNVDEAFEQLVRAVRKYQEQELPPSPPSAPRKKGGGCPCVLL
RUO
RRAS
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers