missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RNF167 Partial ORF (NP_056343, 78 a.a. - 172 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_056343 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 26001 |
Molecular Weight (g/mol) | 35.97kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16167992
|
Abnova™
H00026001-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16157992
|
Abnova™
H00026001-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
RNF167 is an E3 ubiquitin ligase that interacts with TSSC5 (SLC22A18; MIM 602631) and, together with UBCH6 (UBE2E1; MIM 602916), facilitates TSSC5 polyubiquitylation (Yamada and Gorbsky, 2006 [PubMed 16314844]).[supplied by OMIM]
Sequence: VNGSVFIALLRRFDCNFDLKVLNAQKAGYGAAVVHNVNSNELLNMVWNSEEIQQQIWIPSVFIGERSSEYLRALFVYEKGARVLLVPDNTFPLGYSpecifications
NP_056343 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.97kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
5730408C10Rik/DKFZp566H073/LP2254/RING105 | |
RNF167 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
26001 | |
RNF167 (Human) Recombinant Protein (Q01) | |
VNGSVFIALLRRFDCNFDLKVLNAQKAGYGAAVVHNVNSNELLNMVWNSEEIQQQIWIPSVFIGERSSEYLRALFVYEKGARVLLVPDNTFPLGY | |
RUO | |
RNF167 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |