missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RNF14 Full-length ORF (NP_899645.1, 1 a.a. - 348 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009604-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193902

  • 528.00€ / 25µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This protein interacts with androgen receptor (AR) and may function as a coactivator that induces AR target gene expression in prostate. A dominant negative mutant of this gene has been demonstrated to inhibit the AR-mediated growth of prostate cancer. This protein also interacts with class III ubiquitin-conjugating enzymes (E2s) and may act as a ubiquitin-ligase (E3) in the ubiquitination of certain nuclear proteins. Five alternatively spliced transcript variants encoding two distinct isoforms have been reported. [provided by RefSeq]

Sequence: MQFLKEETLAYLNIVSPFELKIGSQKKVQRRTAQASPNTELDFGGAAGSDVDQEEIVDERAVQDVESLSNLIQEILDFDQAQQIKCFNSKLFLCSICFCEKLGSECMYFLECRHVYCKACLKDYFEIQIRDGQVQCLNCPEPKCPSVATPGQVKELVEAELFARYDRLLLQSSLDLMADVVYCPRPCCQLPVMQEPGCTMGICSSCNFAFCTLCRLTYHGVSPCKVTAEKLMDLRNEYLQADEANKRLLDQRYGKRVIQKALEEMESKEWLEKNSKSCPCCGTPIEKLDGCNKMTCTGCMQYFCWICMGSLSRANPYKHFNDPGSPCFNRLFYAVDVDDDIWEDEVED
Specifications

Specifications

NP_899645.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
66kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARA54/FLJ26004/HFB30/HRIHFB2038/TRIAD2
RNF14
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9604
RNF14 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQFLKEETLAYLNIVSPFELKIGSQKKVQRRTAQASPNTELDFGGAAGSDVDQEEIVDERAVQDVESLSNLIQEILDFDQAQQIKCFNSKLFLCSICFCEKLGSECMYFLECRHVYCKACLKDYFEIQIRDGQVQCLNCPEPKCPSVATPGQVKELVEAELFARYDRLLLQSSLDLMADVVYCPRPCCQLPVMQEPGCTMGICSSCNFAFCTLCRLTYHGVSPCKVTAEKLMDLRNEYLQADEANKRLLDQRYGKRVIQKALEEMESKEWLEKNSKSCPCCGTPIEKLDGCNKMTCTGCMQYFCWICMGSLSRANPYKHFNDPGSPCFNRLFYAVDVDDDIWEDEVED
RUO
RNF14
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers