missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RNF12 Full-length ORF (NP_057204.2, 1 a.a. - 624 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_057204.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51132 |
Molecular Weight (g/mol) | 94.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16119682
|
Abnova™
H00051132-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16109682
|
Abnova™
H00051132-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is a RING-H2 zinc finger protein. It has been shown to be an E3 ubiquitin protein ligase that targets LIM domain binding 1 (LDB1/CLIM), and causes proteasome-dependent degradation of LDB1. This protein and LDB1 are co-repressors of LHX1/LIM-1, a homeodomain transcription factor. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]
Sequence: MENSDSNDKGSGDQSAAQRRSQMDRLDREEAFYQFVNNLSEEDYRLMRDNNLLGTPGESTEEELLRRLQQIKEGPPPQNSDENRGGDSSDDVSNGDSIIDWLNSVRQTGNTTRSGQRGNQSWRAVSRTNPNSGDFRFSLEINVNRNNGSQNSENENEPSARRSSGENVENNSQRQVENPRSESTSARPSRSERNSTEALTEVPPTRGQRRARSRSPDHRRTRARAERSRSPLHPMSEIPRRSHHSISSQTFEHPLVNETEGSSRTRHHVTLRQQISGPELLSRGLFAASGTRNASQGAGSSDTAASGESTGSGQRPPTIVLDLQVRRVRPGEYRQRDSIASRTRSRSQTPNNTVTYESERGGFRRTFSRSERAGVRTYVSTIRIPIRRILNTGLSETTSVAIQTMLRQIMTGFGELSYFMYSDSDSEPTGSVSNRNMERAESRSGRGGSGGGSSSGSSSSSSSSSSSSSSSSSSSSPSSSSGGESSETSSDLFEGSNEGSSSSGSSGARREGRHRAPVTFDESGSLPFLSLAQFFLLNEDDDDQPRGLTKEQIDNLAMRSFGENDALKTCSVCITEYTEGNKLRKLPCSHEYHVHCIDRWLSENSTCPICRRAVLASGNRESVVSpecifications
NP_057204.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
94.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MENSDSNDKGSGDQSAAQRRSQMDRLDREEAFYQFVNNLSEEDYRLMRDNNLLGTPGESTEEELLRRLQQIKEGPPPQNSDENRGGDSSDDVSNGDSIIDWLNSVRQTGNTTRSGQRGNQSWRAVSRTNPNSGDFRFSLEINVNRNNGSQNSENENEPSARRSSGENVENNSQRQVENPRSESTSARPSRSERNSTEALTEVPPTRGQRRARSRSPDHRRTRARAERSRSPLHPMSEIPRRSHHSISSQTFEHPLVNETEGSSRTRHHVTLRQQISGPELLSRGLFAASGTRNASQGAGSSDTAASGESTGSGQRPPTIVLDLQVRRVRPGEYRQRDSIASRTRSRSQTPNNTVTYESERGGFRRTFSRSERAGVRTYVSTIRIPIRRILNTGLSETTSVAIQTMLRQIMTGFGELSYFMYSDSDSEPTGSVSNRNMERAESRSGRGGSGGGSSSGSSSSSSSSSSSSSSSSSSSSPSSSSGGESSETSSDLFEGSNEGSSSSGSSGARREGRHRAPVTFDESGSLPFLSLAQFFLLNEDDDDQPRGLTKEQIDNLAMRSFGENDALKTCSVCITEYTEGNKLRKLPCSHEYHVHCIDRWLSENSTCPICRRAVLASGNRESVV | |
RUO | |
RNF12 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
51132 | |
RNF12 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC15161/NY-REN-43/RLIM | |
RNF12 | |
Recombinant | |
wheat germ expression system |