missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RIOK1 Partial ORF (NP_113668, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00083732-Q01.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16025705

  • 341.70€ / 10µg

Estimated Shipment: 24-01-2022
Add to basket

Description

Description

This gene includes two alternatively spliced transcript variants, which encode different isoforms. The function of this gene has not been determined. [provided by RefSeq]

Sequence: MDYRRLLMSRVVPGQFDDADSSDSENRDLKTVKEKDDILFEDLQDNVNENGEGEIEDEEEEGYDDDDDDWDWDEGVGKLAKGYVWNGGSNPQANRQTSDS
Specifications

Specifications

NP_113668
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MDYRRLLMSRVVPGQFDDADSSDSENRDLKTVKEKDDILFEDLQDNVNENGEGEIEDEEEEGYDDDDDDWDWDEGVGKLAKGYVWNGGSNPQANRQTSDS
RUO
RIOK1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
83732
RIOK1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AD034/FLJ30006/MGC12903/bA288G3.1
RIOK1
Yes
Videos
SDS
Documents
Certificates
Special Offers

Special Offers