missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RGS19 Partial ORF (NP_005864, 51 a.a. - 140 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_005864 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10287 |
Molecular Weight (g/mol) | 35.64kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16114832
|
Abnova™
H00010287-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16104832
|
Abnova™
H00010287-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
G proteins mediate a number of cellular processes. The protein encoded by this gene belongs to the RGS (regulators of G-protein signaling) family and specifically interacts with G protein, GAI3. This protein is a guanosine triphosphatase-activating protein that functions to down-regulate Galpha i/Galpha q-linked signaling. Alternatively spliced transcript variants encoding the same protein isoform have been found for this gene. [provided by RefSeq]
Sequence: WNQERRRAWQASRESKLQPLPSCEVCATPSPEEVQSWAQSFDKLMHSPAGRSVFRAFLRTEYSEENMLFWLACEELKAEANQHVVDEKARSpecifications
NP_005864 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GAIP/RGSGAIP | |
RGS19 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10287 | |
RGS19 (Human) Recombinant Protein (Q01) | |
WNQERRRAWQASRESKLQPLPSCEVCATPSPEEVQSWAQSFDKLMHSPAGRSVFRAFLRTEYSEENMLFWLACEELKAEANQHVVDEKAR | |
RUO | |
RGS19 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |