Learn More
Abnova™ Human RFC3 Full-length ORF (AAH00149.1, 1 a.a. - 356 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH00149.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5983 |
Molecular Weight (g/mol) | 64.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16100182
|
Abnova™
H00005983-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16190172
|
Abnova™
H00005983-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kDa. This gene encodes the 38 kDa subunit. This subunit is essential for the interaction between the 140 kDa subunit and the core complex that consists of the 36, 37, and 40 kDa subunits. Alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq]
Sequence: MSLWVDKYRPCSLGRLDYHKEQAAQLRNLVQCGDFPHLLVYGPSGAGKKTRIMCILRELYGVGVEKLRIEHQTITTPSKKKIEISTIASNYHLEVNPSDAGNSDRVVIQEMLKTVAQSQQLETNSQGDFKVVLLTEVDKLTKDAQHALRRTMEKYMSTCRLILCCNSTSKVIPPIRSRCLAVRVPAPSIEDICHVLSTVCKKEGLNLPSQLAHRLAEKSCRNLRKALLMCEACRVQQYPFTADQEIPETDWEVYLRETANAIVSQQTPQRLLEVRGRLYELLTHCIPPEIIMKGLLSELLHNCGGQLKGEVAQMAAYYEHRLQLGSKAIYHLEAFVAKFMALYKKFMEDGLEGMMFSpecifications
AAH00149.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSLWVDKYRPCSLGRLDYHKEQAAQLRNLVQCGDFPHLLVYGPSGAGKKTRIMCILRELYGVGVEKLRIEHQTITTPSKKKIEISTIASNYHLEVNPSDAGNSDRVVIQEMLKTVAQSQQLETNSQGDFKVVLLTEVDKLTKDAQHALRRTMEKYMSTCRLILCCNSTSKVIPPIRSRCLAVRVPAPSIEDICHVLSTVCKKEGLNLPSQLAHRLAEKSCRNLRKALLMCEACRVQQYPFTADQEIPETDWEVYLRETANAIVSQQTPQRLLEVRGRLYELLTHCIPPEIIMKGLLSELLHNCGGQLKGEVAQMAAYYEHRLQLGSKAIYHLEAFVAKFMALYKKFMEDGLEGMMF | |
RUO | |
RFC3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
5983 | |
RFC3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC5276/RFC38 | |
RFC3 | |
Recombinant | |
wheat germ expression system |