missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human REG1A Full-length ORF (NP_002900.2, 1 a.a. - 166 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifications

Accession Number NP_002900.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5967
Molecular Weight (g/mol) 45.1kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16112218
View Documents Promotion Details
Abnova™
H00005967-P02.25UG
25 ug
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
16102218
View Documents Promotion Details
Abnova™
H00005967-P02.10UG
10 ug
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

This gene is a type I subclass member of the Reg gene family. The Reg gene family is a multigene family grouped into four subclasses, types I, II, III and IV, based on the primary structures of the encoded proteins. This gene encodes a protein that is secreted by the exocrine pancreas. It is associated with islet cell regeneration and diabetogenesis and may be involved in pancreatic lithogenesis. Reg family members REG1B, REGL, PAP and this gene are tandemly clustered on chromosome 2p12 and may have arisen from the same ancestral gene by gene duplication. [provided by RefSeq]

Sequence: MAQTSSYFMLISCLMFLSQSQGQEAQTELPQARISCPEGTNAYRSYCYYFNEDRETWVDADLYCQNMNSGNLVSVLTQAEGAFVASLIKESGTDDFNVWIGLHDPKKNRRWHWSSGSLVSYKSWGIGAPSSVNPGYCVSLTSSTGFQKWKDVPCEDKFSFVCKFKN
Specifications

Specifications

NP_002900.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
45.1kDa
Glutathione Sepharose 4 Fast Flow
MAQTSSYFMLISCLMFLSQSQGQEAQTELPQARISCPEGTNAYRSYCYYFNEDRETWVDADLYCQNMNSGNLVSVLTQAEGAFVASLIKESGTDDFNVWIGLHDPKKNRRWHWSSGSLVSYKSWGIGAPSSVNPGYCVSLTSSTGFQKWKDVPCEDKFSFVCKFKN
RUO
REG1A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
5967
REG1A (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ICRF/MGC12447/P19/PSP/PSPS/PSPS1/PTP/REG
REG1A
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications