missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RBM23 Full-length ORF (NP_060577.2, 1 a.a. - 424 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_060577.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55147 |
Molecular Weight (g/mol) | 73.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16131183
|
Abnova™
H00055147-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16121183
|
Abnova™
H00055147-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the U2AF-like family of RNA binding proteins. This protein interacts with some steroid nuclear receptors, localizes to the promoter of a steroid- responsive gene, and increases transcription of steroid-responsive transcriptional reporters in a hormone-dependent manner. It is also implicated in the steroid receptor-dependent regulation of alternative splicing. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MASDDFDIVIEAMLEAPYKKEEDEQQRKEVKKDYPSNTTSSTSNSGNETSGSSTIGETSNRSRDRDRYRRRNSRSRSPGRQCRHRSRSWDRRHGSESRSRDHRREDRVHYRSPPLATGYRYGHSKSPHFREKSPVREPVDNLSPEERDARTVFCMQLAARIRPRDLEDFFSAVGKVRDVRIISDRNSRRSKGIAYVEFCEIQSVPLAIGLTGQRLLGVPIIVQASQAEKNRLAAMANNLQKGNGGPMRLYVGSLHFNITEDMLRGIFEPFGKIDNIVLMKDSDTGRSKGYGFITFSDSECARRALEQLNGFELAGRPMRVGHVTERLDGGTDITFPDGDQELDLGSAGGRFQLMAKLAEGAGIQLPSTAAAAAAAAAAQAAALQLNGAVPLGALNPAALTALSPALNLASQCFQLSSLFTPQTMSpecifications
NP_060577.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MASDDFDIVIEAMLEAPYKKEEDEQQRKEVKKDYPSNTTSSTSNSGNETSGSSTIGETSNRSRDRDRYRRRNSRSRSPGRQCRHRSRSWDRRHGSESRSRDHRREDRVHYRSPPLATGYRYGHSKSPHFREKSPVREPVDNLSPEERDARTVFCMQLAARIRPRDLEDFFSAVGKVRDVRIISDRNSRRSKGIAYVEFCEIQSVPLAIGLTGQRLLGVPIIVQASQAEKNRLAAMANNLQKGNGGPMRLYVGSLHFNITEDMLRGIFEPFGKIDNIVLMKDSDTGRSKGYGFITFSDSECARRALEQLNGFELAGRPMRVGHVTERLDGGTDITFPDGDQELDLGSAGGRFQLMAKLAEGAGIQLPSTAAAAAAAAAAQAAALQLNGAVPLGALNPAALTALSPALNLASQCFQLSSLFTPQTM | |
RUO | |
RBM23 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
55147 | |
RBM23 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAPERbeta/FLJ10482/MGC4458/PP239/RNPC4 | |
RBM23 | |
Recombinant | |
wheat germ expression system |